Få mere at vide
Tocris Bioscience™ GSK 329
Beskrivelse
GSK 329 is a potent and selective TNNI3K (cardiac troponin I-interacting kinase) inhibitor (IC50 = 10 nM). It exhibits selectivity for TNNI3K over human VEGFR2 (40-fold), p38α (80-fold), B-Raf (>200-fold) and displays >100-fold selectivity over 80% of 185 kinases tested. In vivo, GSK 329 reduces infarct size, ROS levels, and p38 activation in a mouse model of ischemia/reperfusion cardiac injury. This compound is orally bioavailable.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-[3,5-Dichloro-4-[[6-(methylamino)-4-pyrimidinyl]oxy]phenyl]-N'-[3-(trifluoromethyl)phenyl]urea |
| CAS | 1268490-12-5 |
| Mængde | 10 mg |
| Mål | MAPK Inhibitors |
| Molekylær formel | C19H14Cl2F3N5O2 |
| Renhed | 0.98 |