Få mere at vide
GSK 321, TRC
Artikelnummer. 30240979
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30240979 1 mg -
30240484 2.5 mg 2.5mg
30240939 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240979 Leverandør Toronto Research Chemicals Leverandørnr. TRCG7973051MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1816331-63-1
Molekylær formel C28H28FN5O3
Molekylvægt (g/mol) 501.55
Synonym (R)-1-(4-Fluorobenzyl)-N-(3-((S)-1-hydroxyethyl)phenyl)-7-methyl-5-(1H-pyrrole-2-carbonyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide, GSK 321
IUPAC navn (7R)-1-[(4-fluorophenyl)methyl]-N-[3-[(1S)-1-hydroxyethyl]phenyl]-7-methyl-5-(1H-pyrrole-2-carbonyl)-6,7-dihydro-4H-pyrazolo[4,3-c]pyridine-3-carboxamide
SMIL C[C@H](O)c1cccc(NC(=O)c2nn(Cc3ccc(F)cc3)c4[C@H](C)CN(Cc24)C(=O)c5ccc[nH]5)c1
Tekniske data
Kemisk navn eller materiale (R)-1-(4-Fluorobenzyl)-N-(3-((S)-1-hydroxyethyl)phenyl)-7-methyl-5-(1H-pyrrole-2-carbonyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide
Mængde 1 mg
InChI formel InChI=1S/C28H28FN5O3/c1-17-14-33(28(37)24-7-4-12-30-24)16-23-25(27(36)31-22-6-3-5-20(13-22)18(2)35)32-34(26(17)23)15-19-8-10-21(29)11-9-19/h3-13,17-18,30,35H,14-16H2,1-2H3,(H,31,36)/t17-,18+/m1/s1
Formel vægt 501.2176