Få mere at vide
Tocris Bioscience™ GSK 2837808A
GREENER_CHOICE
Potent and selective LDHA inhibitor
Brand:  Tocris Bioscience™ 5189/10
Encompass
Produktkode 15776556
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
GSK 2837808A is a potent and selective lactate dehydrogenase A and B (LDHA and LDHB) inhibitor (IC50 values are 2.6 and 43 nM for LDHA and LDHB respectively). Inhibits lactate production in selected cancer cell lines. Reduces glucose uptake and enhances mitochondrial oxygen consumption in Snu398 hepatocellular carcinoma cells. Inhibits proliferation and induces apoptosis in Snu398 cells. Inhibits transcription of histone 2B (H2B) gene in HCT116 and NCM460 cells. Cell permeable.
Tekniske data
Tekniske data
98%
1445879-21-9
10 mg
3-[[3-[(Cyclopropylamino)sulfonyl]-7-(2,4-dimethoxy-5-pyrimidinyl)-4-quinolinyl]amino]-5-(3,5-difluorophenoxy)benzoic acid
-20°C
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only