Få mere at vide
GSK 2194069, Tocris Bioscience™
Potent human fatty acid synthase (hFASN) inhibitor
Brand: Tocris Bioscience 5303/25
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
GSK 2194069 is a potent human fatty acid synthase (hFASN) inhibitor (IC50 = 7.7 nM); inhibits hFAS β-ketoacyl reductase activity. Inhibits lipid synthesis and attenuates proliferation of A549 non-small-cell lung cancer cells (EC50 = 15 nM) in vitro.Tekniske data
| 4-[4-(5-Benzofuranyl)phenyl]-5-[[(3S)-1-(cyclopropylcarbonyl)-3-pyrrolidinyl]methyl]-2,4-dihydro-3H-1,2,4-triazol-3-one | |
| 1332331-08-4 | |
| >99% | |
| 25 mg |