Få mere at vide
GSK 2194069, Tocris Bioscience™
GREENER_CHOICE
Potent human fatty acid synthase (hFASN) inhibitor
Brand:  Tocris Bioscience 5303/5
Encompass
Produktkode 15933221
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
GSK 2194069 is a potent human fatty acid synthase (hFASN) inhibitor (IC50 = 7.7 nM); inhibits hFAS β-ketoacyl reductase activity. Inhibits lipid synthesis and attenuates proliferation of A549 non-small-cell lung cancer cells (EC50 = 15 nM) in vitro.
Tekniske data
Tekniske data
4-[4-(5-Benzofuranyl)phenyl]-5-[[(3S)-1-(cyclopropylcarbonyl)-3-pyrrolidinyl]methyl]-2,4-dihydro-3H-1,2,4-triazol-3-one
1332331-08-4
>99%
5 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner