missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
- Home
- Products
- Personligt beskyttelsesudstyr
- Håndbeskyttelse
- Dry Box Handsker
- AL Glove Systems GSB Isolator Box Gloves
AL Glove Systems GSB Isolator Box Gloves
Artikelnummer. 31337462
Skift visning
Klik for at se tilgængelige muligheder
Størrelse:
Large, 9.5
Medium, 8
X-Large, 11
Længde (metrisk):
800 mm
920 mm
Tykkelse (metrisk):
0.4 mm
0.6 mm
Diameter (metrisk) manchet:
160 mm
180 mm
200 mm
225 mm
250 mm
300 mm
31 product options available for selection
| Artikelnummer. | Størrelse | Længde (metrisk) | Tykkelse (metrisk) | Diameter (metrisk) manchet |
|---|---|---|---|---|
| 31337462 | X-Large, 11 | 920 mm | 0.6 mm | 300 mm |
| 31337400 | Medium, 8 | 800 mm | 0.4 mm | 180 mm |
| 31337436 | Medium, 8 | 800 mm | 0.6 mm | 180 mm |
| 31337484 | Medium, 8 | 800 mm | 0.4 mm | 200 mm |
| 31337398 | Medium, 8 | 800 mm | 0.6 mm | 200 mm |
| 31337454 | Medium, 8 | 800 mm | 0.4 mm | 225 mm |
| 31337483 | Medium, 8 | 800 mm | 0.6 mm | 225 mm |
| 31337482 | Large, 9.5 | 800 mm | 0.4 mm | 160 mm |
| 31337497 | Large, 9.5 | 800 mm | 0.6 mm | 160 mm |
| 31337429 | Large, 9.5 | 800 mm | 0.4 mm | 180 mm |
| 31337439 | Large, 9.5 | 800 mm | 0.6 mm | 200 mm |
| 31337416 | Large, 9.5 | 800 mm | 0.4 mm | 225 mm |
| 31337517 | Large, 9.5 | 800 mm | 0.4 mm | 250 mm |
| 31337495 | Large, 9.5 | 800 mm | 0.6 mm | 250 mm |
| 31337424 | Large, 9.5 | 800 mm | 0.4 mm | 300 mm |
| 31337508 | Large, 9.5 | 800 mm | 0.6 mm | 300 mm |
| 31337461 | Large, 9.5 | 920 mm | 0.4 mm | 200 mm |
| 31337476 | Large, 9.5 | 920 mm | 0.6 mm | 200 mm |
| 31337494 | Large, 9.5 | 920 mm | 0.4 mm | 250 mm |
| 31337469 | Large, 9.5 | 920 mm | 0.6 mm | 250 mm |
| 31337396 | X-Large, 11 | 800 mm | 0.6 mm | 200 mm |
| 31337407 | X-Large, 11 | 800 mm | 0.4 mm | 225 mm |
| 31337493 | X-Large, 11 | 800 mm | 0.4 mm | 250 mm |
| 31337419 | X-Large, 11 | 800 mm | 0.6 mm | 250 mm |
| 31337401 | X-Large, 11 | 800 mm | 0.4 mm | 300 mm |
| 31337457 | X-Large, 11 | 920 mm | 0.4 mm | 300 mm |
| 31337518 | X-Large, 11 | 800 mm | 0.6 mm | 300 mm |
| 31337399 | X-Large, 11 | 920 mm | 0.4 mm | 225 mm |
| 31337467 | X-Large, 11 | 920 mm | 0.6 mm | 225 mm |
| 31337447 | X-Large, 11 | 920 mm | 0.4 mm | 250 mm |
| 31337488 | X-Large, 11 | 920 mm | 0.6 mm | 250 mm |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
Denne vare kan ikke returneres.
Se returpolicy
Artikelnummer.
31337462
Leverandør
AL Glove Systems
Leverandørnr.
1230069111
Log ind eller registrer dig for at se din pris og tilgængelighed.
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
AL Glove Systems GSB gloves are made of bromo-butyl rubber (BBR) with high gas impermeability and high resistance to a wide range of polar hydrocarbons such as acids, esters, ketones, and amine derivatives. They are highly flexible with a high sensitivity. The GSB Glove is classified as PAH-free.
- Impermeable to gas
- High resistance to the permeation of mainly oxygen, nitrogen, inert gases, etc. during production under inert gas atmosphere in inert boxes for the production of electronic components and microcomponents
Flexible:
- Offers good sensitivity in handling
- Resistant to chemicals
- High resistance to a wide range of chemicals and toxins
PERMEATION EN ISO 374-1:2016/A1:2018
- A Methanol Protection Level 6
- B Acetone Protection Level 6
- I Ethyl acetate Protection Level 3
- K Sodium hydroxide 40% Protection Level 6
- L Sulphuric acid 96% Protection Level 6
- N Acetic acid 99% Protection Level 6
- O Ammonium hydroxide 25% Protection Level 6
- T Formaldehyde 37% Protection Level 6
Tekniske data
Tekniske data
| Certificeringer/overholdelse (Europa) | PPE Category III Directive 2016/425 EC Type |
| Kloreret | No |
| Farve | Black |
| Hånd | Ambidextrous |
| Latex fri | Yes |
| Materiale | Butyl |
| Produkttype | Dry Box, Isolator Gloves |
| Størrelse | X-Large, 11 |
| Temperatur (engelsk) Max. | 194°F |
| Temperatur (metrisk) Maks | 90°C |
| Tykkelse (engelsk) | 24 mil |
| Tykkelse (metrisk) | 0.6 mm |
| Til brug med (applikation) | Handling Sensitive or Hazardous Materials in an Inert or Contained Atmosphere |
| Til brug med (udstyr) | Dry Box, Isolator, RABS |
| Greb design | Smooth |
| Revisionsstandarder for fremstilling/QMS | ISO 9001:2015 |
| Emballagetype | Carton |
| Temperatur (engelsk) Min | -40°F |
| Temperatur (metrisk) Min. | -40°C |
| Længde (engelsk) | 36 in. |
| Længde (metrisk) | 920 mm |
| Diameter (engelsk) manchet | 12 In. |
| Diameter (metrisk) manchet | 300 mm |
| Vis mere Vis mindre |
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?