Få mere at vide
Tocris Bioscience™ GS 143
β-TrCP1 ligase inhibitor
Brand: Tocris Bioscience™ 5636/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
GS 143 is a β-TrCP1 ligase inhibitor. Inhibits IκBα ubiquitination (IC50 = 5.2 μM) without affecting proteasome activity. Blocks LPS-induced expression of inflammatory cytokines in human myelomonocytic cells. Suppresses antigen-induced NFκB expression, inflammation and mucus production in airways of OVA-sensitized mice.Tekniske data
| 4-[4-[[5-(2-Fluorophenyl)-2-furanyl]methylene]-4,5-dihydro-5-oxo-3-(phenylmethyl)-1H-pyrazol-1-yl]benzoic acid | |
| 916232-21-8 | |
| 50 mg |