missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Growth Hormone Antibody (GH/4886R), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-14007AF647
Additional Details : Weight : 0.00970kg
Description
Growth Hormone Monoclonal antibody specifically detects Growth Hormone in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Growth Hormone | |
Monoclonal | |
Alexa Fluor 647 | |
GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N | |
Recombinant full-length human Growth Hormone protein (Uniprot: P01241 ) | |
0.1 mL | |
Biologically Active Proteins | |
2688 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
GH/4886R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |