Learn More
GRM4 Mouse anti-Human, Clone: OTI6D1, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810384S
Encompass_Preferred
Product Code. 17583434
  • 1805.36 DKK / 30µL
This item is not returnable. View return policy
Description
Description
GRM4 Monoclonal antibody specifically detects GRM4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
GRM4 Mouse anti-Human, Clone: OTI6D1, TrueMAB™, Origene