Learn More
Fritsch™ Grinding Sets
Brand: Fritsch™ 46.2110.00
Additional Details : Weight : 5.00000kg
Specifications
Grinding Set | |
Mortar Grinder Pulverisette 2 | |
Hard Porcelain |
Soft, fibrous samples | |
Highly centralized mortar bowl rimmed in a plastic frame, pestle, and scraper |