Learn More
SPEX Certiprep™ Grinding cylinder Geno/Grinder Ceramic
Useful for mixing samples for extraction methods such as pesticide residue analysis or extracting oil from fish and seafood.
Brand:  SPEX Certiprep™ 2185
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15350432
  • 605.00 DKK / Pack of 100
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
  • Chemically inert
  • Angle-cut end help shear the sample matrix and ensure good homogenisation and maximise extraction rates
Specifications
Specifications
5 or 15 mL
5/32 x 5/16 in. (4 x 8 mm)
Two cylinders are used per sample vial
100/bag