missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Granzyme M Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553345
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Immunogen sequence: PSMVCLAADS KDQAPCKGDS GGPLVCGKGR VLAGVLSFSS RVCTDIFKPP VATAVAPYVS WIRKVTGR Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 78%.
Cleaves peptide substrates after methionine, leucine, and norleucine. Physiological substrates include EZR, alpha- tubulins and the apoptosis inhibitor BIRC5/Survivin. Promotes caspase activation and subsequent apoptosis of target cells.
Tekniske data
| Granzyme M | |
| Polyclonal | |
| Unconjugated | |
| GZMM | |
| granzyme M; granzyme M (lymphocyte met-ase 1); GZMM; Hu-Met-1; Lmet1; lymphoctye Met-ase 1; lymphocyte met-ase 1; MET1; Met-1 serine protease; Met-ase; MMET-1; natural killer cell granular protease; RNK-Met-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3004 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51124 | |
| GZMM | |
| Recombinant protein corresponding to Human GZMM. Recombinant protein control fragment (Product #RP-90596). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion