Få mere at vide
Granzyme M Mouse anti-Human , Alexa Fluor 647, Clone: 4B2G4, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566996
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Granzyme M Monoclonal antibody specifically detects Granzyme M in Human samples. It is validated for Flow Cytometry
Tekniske data
| Granzyme M | |
| Monoclonal | |
| Alexa Fluor 647 | |
| GZMM; GrM; Gzm M; GzmM; HU-Met-1; met-ase | |
| Granzyme M | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 4B2G4 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed. |