Få mere at vide
Granisetron-d3, TRC
Artikelnummer. 30240540
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30240540 25 mg -
30240559 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30240540 Leverandør Toronto Research Chemicals Leverandørnr. TRCG78000325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1224925-76-1
Molekylær formel C18 D3 H21 N4 O
Molekylvægt (g/mol) 315.43
Synonym BRL 43694-d3, BRL 43964-d3, Granisetron-d3, Kevatril-d3, endo-Granisetron-d3
IUPAC navn N-[(1R,5S)-9-methyl-9-azabicyclo[3.3.1]nonan-3-yl]-1-(trideuteriomethyl)indazole-3-carboxamide
SMIL [2H]C([2H])([2H])n1nc(C(=O)NC2C[C@H]3CCC[C@@H](C2)N3C)c4ccccc14
Tekniske data
Kemisk navn eller materiale 1-(Methyl-d3)-N-[(3-endo)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-Indazole-3-carboxamide
Mængde 25 mg
InChI formel InChI=1S/C18H24N4O/c1-21-13-6-5-7-14(21)11-12(10-13)19-18(23)17-15-8-3-4-9-16(15)22(2)20-17/h3-4,8-9,12-14H,5-7,10-11H2,1-2H3,(H,19,23)/t12-,13+,14-/i2D3
Formel vægt 315.2138