Få mere at vide
Kartell SpA Graduated Wash Bottle
Beskrivelse
Wide mouth for easy, safe filling. Color coded caps for easy identification. Graduated. Ingenious cap designed to avoid involuntary dripping due to pressure build up in bottle - where toxic or radioactive material is being used
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 250 mL |
| Kapacitet (engelsk) | 8.45 oz. |
| Farve | Yellow |
| Materiale | Polyethylene |
| Sterilitet | Non-sterile |
| Lukning inkluderet | Yes |
| Engangs | Reusable |
| Uddannet | Yes |
| Mund | Wide, Threaded |
| Form | Round |
| Vis mere |