Learn More
Fisherbrand™ Graduated AC Round Bottle with Phenolic PolyCone Lined Cap
Catalog Number 15717569
Click to view available options
No. per Case:
48/Cs.
488/Cs.
Multipurpose containers for use with biological and pathological specimens.
  • Used for storing industrial laboratory chemicals and reagents
  • Designed with slight shoulder for easy pouring, handling
  • Facilitates easy mixing, handling, transfer of contents
  • Graduated in both milliliters and ounces
Closure Liner Polycone
Closure Material Phenolic
Closure Type Screw Cap
Product Type Graduated Bottle
Capacity (Metric) 60 mL
Capacity (English) 2 oz.
For Use With (Application) Storing dry materials
Material Glass
No. per Case 48/Cs.
Closure Color Black
Neck Finish 38-400
Closure Included Yes