Learn More
Fisherbrand™ Grade 1107 Cellulose Technical Filter Paper: Discs
Used for filtration of semi fine particles
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11760046
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Thick smooth paper with medium pore structure
Specifications
Specifications
Disc
20 to 30 μm
For Serum, Culture Medium, dyes, tinctures, vegetal extracts, turbid liquids, juices, Eaux de Cologne, perfumes, essences, hair tonics
100 Pack
150 mm
Cellulose
20 to 30 μm
1107
0.38 mm
150 mm