Få mere at vide
GR 203040, Tocris Bioscience™
Potent, selective NK1 receptor antagonist
Brand: Tocris Bioscience 4647/25
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
GR 203040 is a potent and selective NK1 receptor antagonist. Potently inhibits substance P binding to NK1 receptors (pKi = 10.1 to 10.5). Displays antiemetic activity in vivo.Tekniske data
| 168398-02-5 | |
| (2S,3S)-N-[[2-Methoxy-5-(1H-tetrazol-1-yl)phenyl]methyl]-2-phenyl-3-piperidinamine dihydrochloride | |
| 25 mg |