Få mere at vide
Tocris Bioscience™ GR 127935 hydrochloride
GREENER_CHOICE
Artikelnummer. 30001259
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30001259 10 mg
30001260 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30001259 Leverandør Tocris Bioscience™ Leverandørnr. 1477/10
Potent and selective 5-HT1B and 5-HT1D antagonist
Potent and selective 5-HT1B/1D receptor antagonist (pKi values are 8.5 for both guinea pig 5-HT1D and rat 5-HT1B receptors). Displays > 100-fold selectivity over 5HT1A, 5-HT2A, 5-HT2C receptors and other receptor types. Centrally active following oral administration.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 148642-42-6
Molekylær formel C29H32ClN5O3
Tekniske data
Kemisk navn eller materiale GR 127935 hydrochloride
Mængde 10 mg
Formel vægt 534.06
Renhed >99%
Molekylvægt (g/mol) 534.057