missing translation for 'onlineSavingsMsg'
Learn More
Gosogliptin, MedChemExpress Product Code.: 16493758

Gosogliptin, MedChemExpress

Product Code. p-200096466
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16493758

Brand: Medchem Express HY10287

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gosogliptin is a potent and selective inhibitor of dipeptidyl peptidase-IV (DPP-IV).

Gosogliptin is a potent and selective inhibitor of dipeptidyl peptidase-IV (DPP-IV).

Specifications

Chemical Name or Material Gosogliptin
CAS 869490-23-3
Color Brownish Yellow
Purity Grade Notes Research
Molecular Formula C17H24F2N6O
Quantity 10 mg
Synonym PF-00734200 PF-734200
SMILES O=C([C@H](C1)NC[C@H]1N2CCN(CC2)C3=NC=CC=N3)N4CC(CC4)(F)F
Molecular Weight (g/mol) 366.41
Formula Weight 366.41
Percent Purity 98.71%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Gosogliptin, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.