Få mere at vide
MedChemExpress Goserelin acetate
Artikelnummer. 16425808
Encompass_Preferred
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
16425808 10 mg
1 options
Artikelnummer. 16425808 Leverandør MedChemExpress Leverandørnr. HY13673A
MedChemExpress Goserelin acetate (ICI-118630 acetate), a decapeptide analogue of gonadotropin-releasing hormone (GnRH/LHRH), functions as a GnRH agonist. Goserelin acetate can be used for the research of breast cancer, epithelial ovarian cancer and prostate cancer.
Tekniske data
Renhed 98.59%
CAS 145781-92-6
Indhold og opbevaring Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Til brug med (applikation) Cancer-programmed cell death
Molekylvægt (g/mol) 1329.46 g/mol
Produkttype Peptides
Mængde 10 mg
Forsendelsestilstand Room temperature
Opløselighed H2O : 100 mg/mL (75.22 mM∣Need ultrasonic) ∣DMSO : ≥ 28 mg/mL (21.06 mM)
Synonym ICI-118630 acetate