missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gold nanoparticles, 100nm, amine-functionalized, 3kDa PEGylated, OD50, 572nm absorption

Brand:  Alfa Aesar™ J67484.&I

Additional Details : Weight : 0.00001kg

Quantity 0.5mL

 View more versions of this product

Product Code. 15485855

  • 1998.73 DKK / 0.50mL


Estimated Shipment
27-10-2020
Add to basket

Specifications

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]

Specifications

Gold nanoparticles
amine-functionalized, 3kDa PEGylated, OD: 50, 572nm absorption
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
[Au]
196.967
CHEBI:30050
100nm
Pink to Red
0.5mL
7440-57-5
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
Liquid
Light sensitive
Videos
SDS Documents