Få mere at vide
Guldætsemiddel, Thermo Scientific Chemicals
Artikelnummer. p-4408724
Mængde:
3.8 L
Pakningsstørrelse:
3.8 L
Artikelnummer. 15492007
Brand: Thermo Scientific Alfa Aesar 044584.=C
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
High purity, low sodium and 0.2 micron filtration system for semiconductor and microelectronic applicationsGold etchant is a selective general-purpose etchant for gold. It is compatible with negative and positive photoresists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Gold etchant
Mængde 3.8 L
Form Væske
MDL nummer MFCD07370792