Få mere at vide
Goat F(ab')2 anti-Rabbit IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Shop alle Bio Techne produkterBeskrivelse
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot, SDS-Page.
Tekniske data
Tekniske data
| Antigen | IgG (H+L) |
| Applikationer | Western Blot, ELISA, Immunohistochemistry, Dot Blot |
| Klassifikation | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugeret | Unconjugated |
| Fortynding | Western Blot 1:2000-1:10000, ELISA 1:20000, Immunohistochemistry 1:1000-1:5000, SDS-Page |
| Formulering | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Værtsarter | Goat |
| Immunogen | Rabbit IgG whole molecule |
| Oprensningsmetode | Affinity Purified |
| Vis mere |