missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Goat F(ab')2 anti-Mouse IgA/IgG/IgM (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB120-5998
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgA/IgG/IgM (H+L) Polyclonal antibody specifically detects IgA/IgG/IgM (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Tekniske data
| IgA/IgG/IgM (H+L) | |
| Polyclonal | |
| Western Blot 1:2000-1:10000, ELISA 1:10000, Immunohistochemistry 1:1000 - 1:5000 | |
| Goat | |
| Multi-step | |
| RUO | |
| Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| 0.125 M Sodium Borate, 0.075 M Sodium Chloride, 0.005 M EDTA, pH 8.0 | |
| Mouse IgG IgA and IgM whole molecule | |
| 0.5 mg | |
| Secondary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion