missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Goat anti-Rat IgA/IgG/IgM (H+L) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-72783-0.5mg
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgA/IgG/IgM (H+L) Polyclonal antibody specifically detects IgA/IgG/IgM (H+L) in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Tekniske data
| IgA/IgG/IgM (H+L) | |
| Polyclonal | |
| Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000 | |
| Goat | |
| Multi-step | |
| RUO | |
| Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| 0.125 M Sodium Borate, 0.075 M Sodium Chloride, 0.005 M EDTA, pH 8.0 | |
| Rat IgG, IgA and IgM whole molecules | |
| 0.5 mg | |
| Secondary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion