missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 680, Invitrogen™

Goat Polyclonal Secondary Antibody

Brand:  Invitrogen 35569

Additional Details : Weight : 1.81440kg

Product Code. 10517364

  • 1280.00 DKK / 500µL
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Product # 35569 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 35569 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. The antibody has been tested by solid-phase adsorbed to ensure minimal cross-reactivity with human, horse, mouse, swine, goat, bovine, and rat serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. DyLight 680 Amax= 682 nm; Emax= 715 nm. Mole Dye/Mole Protein Ratio is lot-dependent.

Rabbit IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG (H+L) Cross-Adsorbed in Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.
Specifications

Specifications

Rabbit IgG (H+L) Cross-Adsorbed
Polyclonal
DyLight 680
Goat
500 μL
Secondary
4° C
IgG
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
1 mg/mL
PBS with 0.02% sodium azide; pH 7.2
Antigen affinity chromatography
RUO
Rabbit
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers