Få mere at vide
Invitrogen™ Goat anti-Rabbit IgG Fc, Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ A78954
Beskrivelse
This goat anti-rabbit IgG Fc secondary antibody has been affinity-purified and shows minimum cross-reactivity to human, mouse and rat serum proteins. These antibodies have been solid phase adsorbed to ensure class specificity and cross adsorbed using human, mouse and rat immunosorbents to remove cross reactive antibodies. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondary antibodies flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g. flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Alexa Fluor dyes are among the most trusted fluorescent dyes available today. Invitrogen™ Alexa Fluor 555 dye is a bright, orange-fluorescent dye with excitation ideally suited to the 555 nm laser line. For stable signal generation in imaging and flow cytometry, Alexa Fluor 555 dye is pH-insensitive over a wide molar range. Probes with high fluorescence quantum yield and high photostability allow detection of low-abundan...
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Tekniske data
| Rabbit IgG Fc, Cross-Adsorbed | |
| Polyclonal | |
| Alexa Fluor 555 | |
| Goat | |
| Purified | |
| RUO | |
| Rabbit | |
| Secondary Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 5mM sodium azide; pH 7.5 | |
| Gamma Immunoglobin Fc region. | |
| 500 μg | |
| Secondary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.