missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Goat anti-Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody, PE
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ M31504
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
For this goat anti-mouse IgM (Heavy chain) antibody, it is reccommended to use a 1:8 working dilution for optimal performance.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Tekniske data
| Mouse IgM (Heavy chain) Cross-Adsorbed | |
| Polyclonal | |
| PBS with sucrose, 4mg/mL BSA and 0.1% sodium azide | |
| Mouse IgM. | |
| 500 μL | |
| Secondary | |
| 4°C | |
| Liquid |
| Flow Cytometry | |
| PE | |
| Goat | |
| Purified | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion