Få mere at vide
Invitrogen™ Goat anti-Mouse IgG2b Secondary Antibody, Alexa Fluor™ 633
Artikelnummer. 10387452
Skift visning
Mængde:
500 μg
Pakningsstørrelse:
Hver
Artikelnummer. Mængde unitSize
10387452 500 μg Hver
1 options
Artikelnummer. 10387452 Leverandør Invitrogen™ Leverandørnr. A21146
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Goat Polyclonal Secondary Antibody
Fluorescence of this long-wavelength Alexa Fluor dye is not visible by looking through a conventional fluorescence microscope. Product will be shipped at Room Temperature.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen Mouse IgG2b
Applikationer Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Koncentration 2 mg/mL
Konjugeret Alexa Fluor 633
Formulering PBS with 5mM sodium azide; pH 7.5
Værtsarter Goat
Immunogen Mouse IgG2b.
Oprensningsmetode Purified
Mængde 500 μg
Regulatorisk status RUO
Primær eller sekundær Secondary
Målarter Mouse
Indhold og opbevaring 4°C, store in dark
Produkttype Secondary Antibody
Form Liquid
Isotype IgG