Få mere at vide
Goat anti-Mouse IgG1 Heavy Chain Secondary Antibody, Biotin, Novus Biologicals™
Artikelnummer. 18232844 Shop alle Bio Techne produkter
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
18232844 1 mL
1 options
Artikelnummer. 18232844 Leverandør Novus Biologicals Leverandørnr. NBP169914B
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Goat Polyclonal Antibody
IgG1 Heavy Chain Polyclonal antibody specifically detects IgG1 Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen IgG1 Heavy Chain
Applikationer Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Klassifikation Polyclonal
Koncentration 1.0 mg/mL
Konjugeret Biotin
Fortynding Western Blot 1:10000- 1:200000, ELISA 1:10000 - 1:200000, Immunohistochemistry 1:250 - 1:2500, Immunocytochemistry/Immunofluorescence 1:100- 1:500
Værtsarter Goat
Immunogen Mouse IgG1
Oprensningsmetode Affinity Purified
Mængde 1 mL
Regulatorisk status RUO
Primær eller sekundær Secondary
Testspecificitet By immunoelectrophoresis and ELISA this antibody reacts specifically with mouse IgG1. Cross reactivity to mouse IgA, IgM, IgG2a, IgG2b, IgG2c, IgG3 and IgE is undetectable. Some hybridoma clones may express aberrant immunoglobulin-related peptides that are improperly recognized by this antibody.
Målarter Mouse
Indhold og opbevaring Store at 4C. Do not freeze.
Isotype IgG
Vis mere Vis mindre
For Research Use Only