missing translation for 'onlineSavingsMsg'
Få mere at vide

Goat anti-Mouse IgG (H+L), Superclonal™ Recombinant Secondary Antibody, AP, Invitrogen™

Artikelnummer. 15421044
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
Pakningsstørrelse:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15421044 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15421044 Leverandør Invitrogen Leverandørnr. A28184

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

Goat Recombinant Polyclonal Secondary Antibody

The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with rabbit, rat, human, bovine, guinea pig, and donkey IgG is observed. Recombit antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombit antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.

Superclonal™ secondary antibodies are developed to be highly specific for both heavy and light chains of IgG. Utilizing the power of recombit technology and multiple phenotypic screens, Superclonal™ antibodies are selected for enhanced specificity to the target species IgG. These recombit secondary antibodies show similar performance as traditional polyclonal secondary antibodies in common immunoassay applications, with minimal cross-reactivity to other commonly-used species. In addition, their recombit animal-free-origin helps ensures lot-to-lot consistency, reducing the need to optimize each lot in relevant assays. Superclonal™ secondary antibodies are designed to allow scientists to achieve the accurate results they need, without some of the concerns that can impact traditional polyclonal secondary antibodies.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Mouse IgG (H+L)
Applikationer ELISA
Klassifikation Recombinant Polyclonal
Koncentration 1 mg/mL
Konjugeret Alkaline Phosphatase (AP)
Formulering PBS with 1mM MgCl2, 1mM ZnCl2, 50% glycerol and 5mM sodium azide
Værtsarter Goat
Immunogen Recombit full-length protein
Oprensningsmetode Size-exclusion - Dialysis
Mængde 1 mg
Regulatorisk status RUO
Primær eller sekundær Secondary
Målarter Mouse
Indhold og opbevaring 4° C, store in dark, DO NOT FREEZE!
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.