missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Goat anti-Mouse IgG (H+L) Secondary Antibody, PE
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ M300041
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Goat anti-Mouse IgG (H+L) Secondary Antibody for Flow, Western Blot
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Tekniske data
| Mouse IgG (H+L) | |
| Polyclonal | |
| PBS with BSA and 0.1% sodium azide | |
| Gamma Immunoglobins Heavy and Light chains. | |
| 500 μL | |
| Secondary | |
| 4°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| PE | |
| Goat | |
| Purified | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion