Få mere at vide
Invitrogen™ Goat anti-Human IgG (H+L) Secondary Antibody, HRP
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ SA510283
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
This antibody reacts with human IgG and with light chains common with other human immunoglobulins as determined by IEP and ELISA techniques. This antibody may cross react with IgG from other species.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Tekniske data
| Human IgG (H+L) | |
| Polyclonal | |
| HRP | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Human | |
| Secondary Antibody | |
| IgG |
| ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| 50mM HEPES with proprietary stabilizer and 0.002% thimerosal | |
| Human IgG h+l. | |
| 1 mg | |
| Secondary | |
| 4°C | |
| Liquid |