Få mere at vide
Invitrogen™ Goat anti-Human IgG (H+L) Secondary Antibody, HRP
Goat Polyclonal Secondary Antibody
Brand:  Invitrogen™ SA510283
Produktkode 17109665
  • 674.00 DKK / 1mg
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
This antibody reacts with human IgG and with light chains common with other human immunoglobulins as determined by IEP and ELISA techniques. This antibody may cross react with IgG from other species.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Tekniske data
Tekniske data
Human IgG (H+L)
Polyclonal
HRP
Goat
Antigen affinity chromatography
RUO
Human
Secondary Antibody
IgG
ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot
1 mg/mL
50mM HEPES with proprietary stabilizer and 0.002% thimerosal
Human IgG h+l.
1 mg
Secondary
4°C
Liquid
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner