missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Goat anti-Chicken IgY (H+L) Secondary Antibody, DyLight 488, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-74780
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgY (H+L) Polyclonal antibody specifically detects IgY (H+L) in Chicken samples. It is validated for Flow Cytometry, Immunomicroscopy
Tekniske data
| IgY (H+L) | |
| Polyclonal | |
| DyLight 488 | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free | |
| Purified Chicken IgG/Y (H&L) | |
| 1 mg | |
| Secondary | |
| Chicken | |
| Purified |
| Flow Cytometry, Immunomicroscopy | |
| LYOPH | |
| Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 | |
| Goat | |
| Affinity purified | |
| RUO | |
| Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
| Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion