missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
STEMCELL Technologies Gö6983
Beskrivelse
- Gö6983 inhibits several isoforms of protein kinase C (PKC; IC50 = 7, 7, 6, 10, 60, and 20,000 nM for PKCα, PKCβ, PKCγ, PKCδ, PKCζ, and PKCμ, respectively) (Gschwendt et al.)
- Enhances reprogramming of mouse embryonic fibroblasts to induced pluripotent stem cells (Dutta et al.)
- Direct lineage reprogramming of fibroblasts to mature neurons, in combination with CHIR99021, RepSox, Forskolin, SP600125, VaLproic Acid, and Y-27632 (Hu et al.)
- Inhibits differentiation and maintains pluripotency in mouse embryonic stem cells (Dutta et al.)
- Enhances human ground state pluripotent stem cell viability and growth (Gafni et al.)
- Inhibits proliferation of human primary fetal bone cells (Krattinger et al.)
- Inhibits the formation of protoplatelets from megakaryocytes derived from adult mouse bone marrow (Williams et al.)
Tekniske data
Tekniske data
| CAS | 133053-19-7 |
| Synonym | Gö 6983 |
| Mængde | 1 mg |
| Molekylær formel | C26H26N4O3 |
| Renhed | ≥98% |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?