missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GnRHR Rabbit anti-Human, Clone: GNRHR/2982R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07747-20UG
Additional Details : Weight : 0.00970kg
Description
GnRHR Monoclonal specifically detects GnRHR in Human samples. It is validated for ELISA.Specifications
GnRHR | |
Monoclonal | |
0.2 mg/ml | |
ELISA | |
GnRH receptor, GnRH-R, GNRHR1, gonadotropin-releasing hormone (type 1) receptor 1, gonadotropin-releasing hormone receptor, GRHR, leutinizing-releasing hormone receptor, LHRHRleutinizing hormone releasing horomone receptor, LRHR, luliberin receptor, type I GnRH receptor | |
Recombinant full-length human GnRHR protein | |
20 μg | |
Primary | |
Human | |
Purified |
ELISA | |
GNRHR/2982R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
2798 | |
Store at 4°C. | |
IgG |