missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNGT1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-00174AF750
Additional Details : Weight : 0.00970kg
Description
GNGT1 Polyclonal specifically detects GNGT1 in Human samples. It is validated for ELISA.Specifications
GNGT1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 750 | |
GNG1, guanine nucleotide binding protein (G protein), gamma transducing activitypolypeptide 1, guanine nucleotide-binding protein G(T) subunit gamma-T1, Transducin gamma chain | |
Produced in rabbits immunized with purified, recombinant GNGT1 (P63211; Pro2-Cys71). | |
0.1 mL | |
Signal Transduction | |
2792 | |
Store at 4°C in the dark. | |
IgG |