Få mere at vide
Tocris Bioscience™ GNF 7
Beskrivelse
GNF 7 is a potent Bcr-Abl inhibitor (IC50 values are 61 and 133 nM for T315I and wild-type Bcr-Abl, respectively), inhibiting a range of clinically relevant Bcr-Abl mutants such as T315I, G250E, E255V, F317L and M351T. GNF 7 is also a Ras signaling inhibitor as well as inhibiting Ack1 and germinal center kinase (GCK). GNF 7 induces cell cycle arrest and apoptosis in leukemia cells harboring NRAS mutations, and shows growth inhibitory activity in human colon cancer cells. It prolongs survival in a leukemia cell xenotransplantation model in mice and is orally bioavailable.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ras GTPase Inhibitors |
| CAS | 839706-07-9 |
| Mængde | 50 mg |
| Mål | Small G Protein Inhibitors |
| Molekylær formel | C28H24F3N7O2 |
| Formel vægt | 547.53 |
| Renhed | >98% |