missing translation for 'onlineSavingsMsg'
Få mere at vide

GNA15 Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12078-1-AP-150UL

 Se flere versioner af dette produkt

Produktkode 30234960

  • 3059.75 DKK / 150µL

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

The GNA15 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GNA15. This antibody recognizes human, mouse, rat antigen. The GNA15 antibody has been validated for the following applications: WB, IHC, ELISA analysis.
Tekniske data

Tekniske data

GNA15
Polyclonal
Unconjugated
Rabbit
Antigen affinity purification
RUO
2769
Store at -20°C. Stable for one year after shipment.
Polyclonal Antibody
IgG
Western Blot, Immunohistochemistry, ELISA
260 μg/mL
GNA16
Recombinant Protein
150 μL
Primary
Human, Mouse, Rat
44 kDa
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.