missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
AcroBiosystems GMP Human Laminin 511 Protein
Beskrivelse
Laminin 511 E8 is a truncated laminin 511, which consists of the C-terminal end of the alpha 5, beta 1, and gamma 1 chains, as one of the ECM proteins. Laminin E8s serve as functionally minimal forms that retain the full capability for binding to integrins. More importantly, laminin 511 E8 is the key matrix for feeder-free culture of human embryonic (hESC) and induced pluripotent stem cells (iPSC). Furthermore, laminin 511 E8 could also support the self-renewal of hESC and iPSC without spontaneous differentiation, in combination with defined stem cell culture media.
Tekniske data
Tekniske data
| Format | Liquid |
| Formulering | Supplied as 0.2 μm filtered solution in PBS, pH 7.4 with protectants. Contact us for customized product form or formulation. |
| Mængde | 10.5 mg |
| Biologisk aktivitet | GMP Human Laminin 511 Protein (Cat. No. GMP-LA1H25) immobilized on CM5 Chip can bind Human Integrin alpha 6 beta 1 (ITGA6&ITGB1) Heterodimer Protein, His Tag&Tag Free with an affinity constant between 1.00 nM - 15.0 nM as determined in a SPR assay (Biacore 8K) (QC tested).∣The SPR based assay shows that GMP Human Laminin 511 Protein (Cat. No. GMP-LA1H25) is stable at 37°C for 23 days.∣The SPR based assay shows that GMP Human Laminin 511 Protein (Cat. No. GMP-LA1H25) is stable after freezing and thawing 3 times.∣The SPR based assay shows batch-to-batch consistency between Acro's GMP and PG Laminin 511.∣GMP Human Laminin 511 Protein (Cat. No. GMP-LA1H25) and Human Laminin 511 Protein, premium grade (Cat. No. LA8-H5283L) could maintain the stemness of iPSC at least Passage 5 and has similar performance. FACS data indicated that the iPSCs expressed high levels of pluripotency associated markers OCT4, SOX2, and SSEA4.∣GMP Human Laminin 511 Protein (Cat. No. GMP-LA1H25) and Human Laminin 511 Protein, premium grade (Cat. No. LA8-H5283L) could maintain the stemness of iPSC at least Passage 5 and have similar performance. Immunofluorescence staining indicated that the iPSCs expressed high levels of pluripotency associated markers OCT4, SOX2, and NANOG. |
| Arter | Human |
| Indhold og opbevaring | 2°C to 8°C for 6 months under sterile conditions; -20°C or below for 3 years. For long term storage, the product should be stored at liquid state at -20°C or below. |
| Forsendelsestilstand | This product is supplied and shipped with dry ice, please inquire the shipping cost. |
| Renhed eller kvalitetsklasse | 0.95 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?