missing translation for 'onlineSavingsMsg'
Få mere at vide

AcroBiosystems GMP Human IL-18 Protein

Artikelnummer. 31452576
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 mg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
31452576 10 mg
31453187 50 μg
31453194 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31452576 Leverandør AcroBiosystems Leverandørnr. GMPL18H1610mg

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

Interleukin-18 (IL-18) is a potent proinflammatory cytokine that induces interferon-gamma (IFN-gamma) production from Th1 cells, NK cells and activated macrophages, particularly in the presence of IL-12. IL-18 also functions in developmental regulation of T-lymphocyte helper type I cells and in Fas-mediated cytotoxicity. Suppression of IL-18 activity is being investigated for treatment of chronic inflammatory diseases such as Crohn's disease and rheumatoid arthritis. It acts by inducing heterodimerization of the two subunits of its receptor, IL-18RAlpha and IL-18RBeta shows structural similarity to IL-1.

Tekniske data

Adgangsnummer Uniprot
Karakterisering IL-18 Structure This protein carries no tag. The protein has a calculated MW of 18.2 kDa. The protein migrates as 17 kDa±3 kDa when calibrated against under reducing (R) condition (SDS-PAGE).
Format Powder-GMP
Formulering Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with protectants. Contact us for customized product form or formulation.
Kvalitetskontrol test <0.5 ng/μg of protein tested by ELISA.∣<0.02 ng/μg of protein tested by qPCR.∣Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.∣Sterility testing was performed using the membrane filtration method in compliance with USP &CM and Ph. Eur. 2.6.1.∣Negative
Mængde 10 mg
Kilde GMP Human IL-18 Protein (GMP-L18H16) is expressed from E. coli cells. It contains AA Tyr 37 - Asp 193 (Accession # ). Request for sequence
Biologisk aktivitet GMP Human IL-18 Protein (Cat. No. GMP-L18H16) stimulates secretion of IFN-γ by KG-1 cells. The specific activity of GMP Human IL-18 Protein is >3.00x10^6 IU/mg, which is calibrated against WHO Reference Reagent Interleukin-18 (Human rDNA derived) (NIBSC code: 03/200) (QC tested).∣The activity of GMP Human IL-18 Protein (Cat. No. GMP-L18H16) was higher than other competing products.∣Human PBMCs were cultured with GMP Human IL-2 Protein (ACROBiosystems, Cat. No. GMP-L02H14), GMP Human IL-18 Protein (ACROBiosystems, Cat. No. GMP-L18H16) and GMP Human 4-1BB Ligand Protein (ACROBiosystems, Cat. No. GMP-41LH26), in CelThreaTM GMP T Cell Expansion Medium (ACROBiosystems, Cat. No. GMP-CM3103) for two weeks. The result shows that GMP Human IL-18 Protein can promote the expansion of these cells with a reasonable cell viability. Notably, the cells exhibit better expansion in GMP Human IL-18 Protein (ACROBiosystems).∣Human PBMCs were cultured with GMP Human IL-2 Protein (ACROBiosystems, Cat. No. GMP-L02H14), GMP Human IL-18 Protein (ACROBiosystems, Cat. No. GMP-L18H16) and GMP Human 4-1BB Ligand Protein (ACROBiosystems, Cat. No. GMP-41LH26), in CelThreaTM GMP T Cell Expansion Medium (ACROBiosystems, Cat. No. GMP-CM3103) for two weeks. The result shows that GMP Human IL-18 Protein (ACROBiosystems) can be comparable with the Competitor M IL-18 protein.∣The Cell based assay shows that GMP Human IL-18 Protein (Cat. No. GMP-L18H16) is stable after freezing and thawing 3 times.∣The Cell based assay shows batch-to-batch consistency between Acro's GMP and PG IL-18.∣MALDI-TOF analysis of GMP Human IL-18 Protein (Cat. No. GMP-L18H16), showing a major peak at 18217.2 Da. The minor peak following the major peak is a matrix-associated artifact of the MALDI-TOF (Routinely tested).
Arter Human
Sterilitet Sterile
Indhold og opbevaring -20°C to -70°C for 5 years in lyophilized state; -70°C for 12 months under sterile conditions after reconstitution. Upon receipt, store it immediately at -20°C or lower for long term storage.
Endotoksin niveau Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.
Forsendelsestilstand This product is supplied and shipped with blue ice, please inquire the shipping cost.
Specifik aktivitet The activity of GMP Human IL-18 Protein (Cat. No. GMP-L18H16) was higher than other competing products.
Renhed eller kvalitetsklasse 0.95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.