missing translation for 'onlineSavingsMsg'
Få mere at vide

AcroBiosystems GMP Human BMP-4 / BMP2B Protein

Artikelnummer. 31453197
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 mg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
31453197 50 μg
31451640 1 mg
31453212 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31453197 Leverandør AcroBiosystems Leverandørnr. GMPBM4H3650ug

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

Bone Morphogenetic Protein 4 (BMP4) is a member of growth factor of the TGF-beta superfamily that plays essential roles in many developmental processes, including neurogenesis, vascular development, angiogenesis and osteogenesis. BMP-4 Initiates the canonical BMP signaling cascade by associating with type I receptor BMPR1A and type II receptor BMPR2. BMP-4 can acts in concert with PTHLH/PTHRP to stimulate ductal outgrowth during embryonic mammary development and to inhibit hair follicle induction.

Tekniske data

Adgangsnummer Uniprot
Karakterisering BMP-4 Structure This protein carries no tag. The protein has a calculated MW of 13.1 kDa. The protein migrates as 21 kDa±3 kDa when calibrated against under reducing (R) condition (SDS-PAGE) due to glycosylation.
Format Powder-GMP
Formulering Lyophilized from 0.22 μm filtered solution in 20 mM Citric acid, pH 2.2 with protectants. Contact us for customized product form or formulation.
Kvalitetskontrol test <0.5 ng/μg of protein tested by ELISA.∣<0.02 ng/μg of protein tested by qPCR.∣Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.∣Sterility testing was performed using the membrane filtration method in compliance with USP &CM and Ph. Eur. 2.6.1.∣Negative
Mængde 50 μg
Kilde GMP Human BMP-4 Protein (GMP-BM4H36) is expressed from CHO cells. It contains AA Ser 293 - Arg 408 (Accession # ). Predicted N-terminus: Ser 293 Request for sequence
Biologisk aktivitet GMP Human BMP-4 Protein (Cat. No. GMP-BM4H36) stimulates Human BMP (Luc) HEK293 Reporter cells. The typical EC50 for this effect is 2.563 ng/mL (QC tested).∣GMP Human BMP-4 Protein (Cat. No. GMP-BM4H36) could promote the differentiation of iPSCs into hematopoietic stem and progenitor cells in a feeder-free system at Day 14, with high expression of CD34+ CD45+. GMP Human BMP-4 Protein (Cat. No. GMP-BM4H36) holds a similar performance with Human BMP-4 Protein, premium grade (Cat. No. BM4-H5317).∣The Cell based assay shows that GMP Human BMP-4 Protein (Cat. No. GMP-BM4H36) is stable at 37°C for 7 hours.∣The Cell based assay shows that GMP Human BMP-4 Protein (Cat. No. GMP-BM4H36) is stable at 4°C for 180 days.∣The Cell based assay shows that GMP Human BMP-4 Protein (Cat. No. GMP-BM4H36) is stable after freezing and thawing 3 times.
Arter Human
Sterilitet Sterile
Indhold og opbevaring -20°C to -70°C for 5 years in lyophilized state; -70°C for 12 months under sterile conditions after reconstitution. Upon receipt, store it immediately at -20°C or lower for long term storage.
Endotoksin niveau Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.
Forsendelsestilstand This product is supplied and shipped with blue ice, please inquire the shipping cost.
Renhed eller kvalitetsklasse 0.95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.