missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ GM-CSF Recombinant Human Protein
Recombinant Human GM-CSF
17570.00 DKK - 48050.00 DKK
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 14 kDa |
Purification Method | Gel-Purified |
Source | Human |
Gene Alias | GM-CSF |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10650111
|
Gibco™
CTP2011 |
100 μg |
17570.00 DKK
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10660111
|
Gibco™
CTP2013 |
1 mg |
48050.00 DKK
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Involved in many biological responses including growth and development of granulocyte and macrophage progenitor cells
- For stimulation and initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils
- ED50 range = 0.02 to 0.1 ng/mL, determined by dose dependent proliferation of human TF-1 cells
- Optimal concentration for individual application should be determined by dose response assay
Specifications
Cell Culture | |
Gel-Purified | |
GM-CSF | |
Recombinant | |
Proliferation of TF1 cells | |
Human | |
E. coli | |
GM-CSF, G-CSF, & M-CSF | |
Gibco™ |
14 kDa | |
Human | |
Human | |
Carrier-Free | |
<0.1 ng/μg | |
Wet Ice | |
Full Length, Recombinant, Ligand | |
Clinical Research, Oncology, Stem Cell Research, Differentiation, Signal Transduction, Drug Development |
For Research Use Only. Not for use in diagnostic procedures.