Learn More
Glyoxylic Acid Monohydrate, Sigma-Aldrich™
Product Code. 15859100
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15859100 250 g 250g
15839100 25 g 25g
15849100 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15859100 Supplier Sigma Aldrich Supplier No. 8041070250
 Restricted Item
Forventet forsendelse: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 563-96-2
Specifications
Melting Point 46.7°C
MDL Number MFCD00149497
Quantity 250 g
Synonym Glyoxylic acid monohydrate, Glyoxylic acid monohydrate
Molecular Weight (g/mol) 92.05
Grade Research