Få mere at vide
Glycyl-Glycyl-Glycine, TRC
Artikelnummer. 30134806
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134806 5 g
30012363 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134806 Leverandør Toronto Research Chemicals Leverandørnr. TRCG7400105G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 556-33-2
Molekylær formel C6 H11 N3 O4
Molekylvægt (g/mol) 189.17
Synonym Glycylglycylglycine, Glycine, N-(N-glycylglycyl)- (6CI,8CI), Diglycylglycine, Glycine tripeptide, Glycylglycylglycine, N-(N'-Glycylglycyl)glycine, NSC 46707, Triglycine
IUPAC navn 2-[[2-[(2-aminoacetyl)amino]acetyl]amino]acetic acid
SMIL NCC(=O)NCC(=O)NCC(=O)O
Tekniske data
Kemisk navn eller materiale Triglycine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C6H11N3O4/c7-1-4(10)8-2-5(11)9-3-6(12)13/h1-3,7H2,(H,8,10)(H,9,11)(H,12,13)
Mængde 5 g
Formel vægt 189.075
Procent renhed >95%