missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Glycopyrronium Bromide, LoGiCal
Specifications
Specifications
| Kemisk navn eller materiale | Glycopyrronium Bromide |
| CAS | 51186-83-5 |
| Analyt- eller komponentnavne | Glycopyrronium Bromide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H28 N O3 . Br |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Glycopyrronium Bromide, Pyrrolidinium, 3-[[(2R)-2-cyclopentyl-2-hydroxy-2-phenylacetyl]oxy]-1,1-dimethyl-, bromide (1:1), (3S)-rel-, Pyrrolidinium, 3-[(cyclopentylhydroxyphenylacetyl)oxy]-1,1-dimethyl-, bromide, (R*,S*)-(±)-, Pyrrolidinium, 3-[[(2R)-cyclopentylhydroxyphenylacetyl]oxy]-1,1-dimethyl-, bromide, (3S)-rel- (9CI), (3S)-rel-3-[[(2R)-2-Cyclopentyl-2-hydroxy-2-phenylacetyl]oxy]-1,1-dimethylpyrrolidinium bromide (1:1), (R*,S*)-3-[(Cyclopentylhydroxyphenylacetyl)oxy]-1,1-dimethylpyrrolidinium bromide, (R*,S*)-(±)-3-[(Cyclopentylhydroxyphenylacetyl)oxy]-1,1-dimethylpyrrolidinium bromide, erythro-Glycopyrronium bromide |
| InChI formel | InChI=1S/C19H28NO3.BrH/c1-20(2)13-12-17(14-20)23-18(21)19(22,16-10-6-7-11-16)15-8-4-3-5-9-15;/h3-5,8-9,16-17,22H,6-7,10-14H2,1-2H3;1H/q+1;/p-1/t17-,19-;/m0./s1 |
| Løsningstype | Neat |
| SMIL | [Br-].C[N+]1(C)CC[C@@H](C1)OC(=O)[C@@](O)(C2CCCC2)c3ccccc3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?