Få mere at vide
Glycolithocholic-2,2,4,4-d4 Acid, TRC
Artikelnummer. 30473217
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30473217 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473217 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46920610MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glycolithocholic-2,2,4,4-d4 Acid
CAS 2044276-16-4
Molekylær formel C26 D4 H39 N O4
Mængde 10 mg
Synonym Glycine, N-[(3α,5β)-3-hydroxy-24-oxocholan-24-yl-2,2,4,4-d4]-, Glycolithocholic acid (2,2,4,4-D4), N-[(3α,5β)-3-Hydroxy-24-oxocholan-24-yl-2,2,4,4-d4]glycine, 3α-Hydroxy-2,2,4,4-tetradeuterio-5β-cholanoylglycine, 3α-Hydroxy-N-(carboxymethyl)-5β-cholan-24-amide-2,2,4,4-d4, Lithocholylglycine-d4, Lithocholylglycine-2,2,4,4-d4
InChI formel InChI=1S/C26H43NO4/c1-16(4-9-23(29)27-15-24(30)31)20-7-8-21-19-6-5-17-14-18(28)10-12-25(17,2)22(19)11-13-26(20,21)3/h16-22,28H,4-15H2,1-3H3,(H,27,29)(H,30,31)/t16-,17-,18-,19+,20-,21+,22+,25+,26-/m1/s1/i10D2,14D2
SMIL [2H]C1([2H])C[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC[C@@H]2C([2H])([2H])[C@@H]1O)[C@H](C)CCC(=O)NCC(=O)O
IUPAC navn 2-[[(4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-2,2,4,4-tetradeuterio-3-hydroxy-10,13-dimethyl-3,5,6,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetic acid
Molekylvægt (g/mol) 437.6486
Formel vægt 437.3443