Learn More
Glycolic acid, Sigma-Aldrich™
Product Code. 32238727
Change view
Click to view available options
Quantity:
25 L
Unit Size:
25 L
Product Code. Quantity Unit Size
32238727 25 L 25 L
1 options
Product Code. 32238727 Supplier Sigma Aldrich Supplier No. 8141869025
 Restricted Item
Forventet forsendelse: 26-11-2026
Add to basket
Add to basket
(70% solution in water) for synthesis
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Glycolic acid
pH 0.1 (25°C in H2O)
Melting Point 10°C
Boiling Point 112°C (1013 hPa)
Density 1.262 g/cm³ at 20°C
Quantity 25 L
Grade Not Available