Få mere at vide
Glycolic-2,2-d2 Acid, TRC
Artikelnummer. 30405288
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30405288 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405288 Leverandør Toronto Research Chemicals Leverandørnr. TRCG64560125MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Glycolic-2,2-d2 acid
CAS 75502-10-2
Renhedsgrad noter HPLC
Molekylær formel C2 D2 H2 O3
Mængde 25 mg
Synonym Acetic-d2 acid, hydroxy- (9CI), Glycolic-2,2-d2 acid (6CI), Glycolic acid-2-2H2, Glycolic acid-d2, 2-Hydroxyacetic-2,2-d2 acid, α-Hydroxyacetic acid-d2, α-Hydroxyacetic-α,α-d2 acid, Glycolic Acid-2,2-D2
InChI formel InChI=1S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)/i1D2
SMIL [2H]C([2H])(O)C(=O)O
IUPAC navn 2,2-dideuterio-2-hydroxy-acetic acid
Molekylvægt (g/mol) 78.06
Formel vægt 78.0286
Procent renhed >95